Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Amoeba Model
SCI_ED

Catalog No. S15643 Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W x H):
9.0 x 6.0 x 2.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S15643 9.0 x 6.0 x 2.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15643 Supplier Eisco™ Supplier No. ZM0014
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Model of Amoeba with organelles exposed.

  • 9.0 x 6.0 x 2.5 in.
  • 1.1 lb.

Specifications

Dimensions (L x W x H) 9.0 x 6.0 x 2.5 in.
Material ABS Plastic
Weight (English) 1.1 lb.
Product Type Amoeba Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.