Learn More
Eisco™ Equal Mass Metal Cylinders Set
SCI_ED
Catalog No. S07850 Shop All Eisco Products
Change view
Click to view available options
Quantity:
Set of 5
Catalog No. Quantity
S07850 Set of 5
1 options
Catalog No. S07850 Supplier Eisco™ Supplier No. PH0112FCB
Add to Cart
Add to Cart
Ideal to study density and specific gravity.
  • Set includes five cylinders of varying length but equal mass
  • Made of zinc, copper, aluminum, tin and lead
Specifications
Quantity Set of 5
For Use With (Application) To study the relationship between mass and density
Product Type Equal Mass Cylinders Set
Includes Set of five cylinder of varying length made of zinc, copper, aluminum, tin and lead
Material Zinc, Copper, Aluminum, Tin and Lead
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov