Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Fulcrum Balance Model
SCI_ED

Catalog No. S07825 Shop All Eisco Products
Change view
Click to view available options
Fulcrum Balance Model
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S07825 Fulcrum Balance Model
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07825 Supplier Eisco™ Supplier No. WDMS17
Add to Cart
Add to Cart

Includes

Activity guide

Illustrates how variations to the fulcrum distance affect the mechanical advantage.

  • Allows students to discover the balance point for a given set of masses
  • Durable wooden construction is ideal for hands-on classroom use

Specifications

Product Type Model, Fulcrum Balance
Material Wood
For Use With (Application) Students can intuitively figure out the balance point for a given set of masses
Includes Activity guide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.