Learn More
Eisco™ Hall's Car Pair
SCI_ED
Catalog No. S96317 Shop All Eisco Products
Change view
Click to view available options
Product Type:
Hall'S Car
Catalog No. Product Type
S96317 Hall'S Car
1 options
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S96317 Supplier Eisco™ Supplier No. PH0351APL
Add to Cart
Add to Cart
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Essential items for dynamics experiments.
Use in conjunction with inclined plane, pulley and weights to study relationship between work and energy.
  • Low-friction wheels
  • Oil-free sleeve bearings — never needs adjustment
  • Durable plastic molded body has deep well for weights and low profile
Specifications
Product Type Hall'S Car
For Use With (Application) For use in dynamics experiments and use to study relationship between work and energy
For Use With (Equipment) Use in conjunction with inclined plane, pulley and weights