Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Hoffmans Brass Clips
SCI_ED

Catalog No. S07859 Shop All Eisco Products
Change view
Click to view available options
Material:
Brass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S07859 Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07859 Supplier Eisco™ Supplier No. CH0298B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For holding or closing various tubing.

  • Nickel plated with projecting plate to prevent damage to tubing by screw
  • Hinged bottom bar for direct fitting and removal without access to ends of tube

Specifications

Type Joint Clip
Material Brass
For Use With (Equipment) Holding or closing various tubing
Length (English) 8.26 in.
Length (Metric) 21 cm
Color Silver
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.