Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Human Embryo Model, 4 weeks old
SCI_ED

Catalog No. S15606 Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W x H):
7.0 x 5.0 x 5.0 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S15606 7.0 x 5.0 x 5.0 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15606 Supplier Eisco™ Supplier No. AM121AAS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Magnified approximately 50 times this model shows the early stages of human development.

Displays the origins of organs and structures. Mounts on a base but can be removed for closer study.

Specifications

Dimensions (L x W x H) 7.0 x 5.0 x 5.0 in.
Includes Numbered with English Key Card
Material ABS Plastic
Mounting Base
Weight (English) 0.7 lb.
Product Type Embryo Model (4 weeks old)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.