Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Human Head, Cross sectional
SCI_ED

Catalog No. S15614 Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W x H):
10.0 x 9.0 x 13.0 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S15614 10.0 x 9.0 x 13.0 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15614 Supplier Eisco™ Supplier No. AM25AS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The model comprehensively showcases the human head and neck muscles, vessels, nerves and internal structures.

The life-size human head and neck model is a very highly detailed map of all of the muscles, vessels, and nerves in the human head and also provides a cross sectional view of the internal structures inside the cranium. Inside of the head, the major anatomy is labeled and colored, allowing students to examine structures such as the brain, mouth, and larynx. The model is mounted on a solid base to allow for decades of student use.

Specifications

Dimensions (L x W x H) 10.0 x 9.0 x 13.0 in.
Includes Numbered with English Key Card
Material PVC
Mounting Base
Weight (English) 0.2 lb.
Product Type Human Head Cross Section Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.