Learn More
Eisco™ Inclined Plane and Cart Model
SCI_ED
Catalog No. S07818 Shop All Eisco Products
Change view
Click to view available options
Inclined Plane and Cart Model
Catalog No.
S07818 Inclined Plane and Cart Model
1 options
Catalog No. S07818 Supplier Eisco™ Supplier No. WDMS10
Add to Cart
Add to Cart
Includes
Small cart, pulley, string, mass and activity guide
Elegantly designed wooden machine for easy student use.
  • Grooves in track for easier use and more consistent results
  • With built-in mass and shallow indent for adding additional mass if needed
  • Allows students to study several different simple machine concepts with one apparatus
Specifications
Material Composite Particle Wood
For Use With (Application) Allows students to study several different simple machine concepts with one apparatus
Includes Small cart, pulley, string, mass and activity guide