Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Iron-Clad Magnet
SCI_ED

Catalog No. S97807 Shop All Eisco Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S97807 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S97807 Supplier Eisco™ Supplier No. PH0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Iron-Clad Magnet is a high power electromagnet capable of lifting up to 100 lb.

  • Precision machine makes contact with the energized part of the magnet
  • 2 lb.

Specifications

For Use With (Application) High power electromagnet capable of lifting up to 100 lb.
Grade 9 to 12
Product Type Iron Clad Electromagnet
Weight (Metric) 0.9 kg
Weight (English) 2 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.