Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Lattice Clamp
SCI_ED

Catalog No. s02608 Shop All Eisco Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S02608 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02608 Supplier Eisco™ Supplier No. 57060250/DEL

For lattice building and rod coupling.

  • Uses 12.5mm dia. rods
  • Fitted with screws for Allen wrench

Specifications

Type Lattice Clamp
Material Glass
For Use With (Equipment) Lattice building and rod coupling
For Use With (Application) Lattice building and rod coupling
Color Silver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.