Learn More
Eisco™ Linear Air Track
SCI_ED
Catalog No. S05827 Shop All Eisco Products
Change view
Click to view available options
Linear Air Track
Catalog No.
S05827 Linear Air Track
1 options
Catalog No. S05827 Supplier Eisco™ Supplier No. PH0362
Add to Cart
Add to Cart
Includes
Comprehensive accessory pack to facilitate a variety of demonstrations
Dramatically demonstrate the field of collisions and conservation of momentum.
Cost-effective, virtually friction-free system for the study of all aspects of dynamics: velocity, force and acceleration, potential energy, kinetic energy, conservation of energy, SHM, motion on level and inclined planes.
  • 2m aluminum track made from strong 2.2 x 2.2 in. (56 x 56mm) aluminum extrusion
  • With leveling feet
  • Gliders designed to accept standard slotted masses for additional loads
  • Recommended operating temperature: 10° to 40°C
  • Recommended operating humidity: <85%
Standard light gates and timers are required for measurement of glider speeds.
Order Info
Overall dimensions: 80.9 x 8.8 x 7.1 in. (205.5 x 22.4 x 18.0cm). Total weight: 19.8 lb. (9kg). Glider length: 4.7 in. (12.0cm), 9.4 in. (24.0cm). Supplied without air blower.
Specifications
Product Type Linear Air Track
Material Aluminum
For Use With (Application) Dramatically demonstrates the field of collisions and conservation of momentum, used for the study of all aspects of dynamics like velocity, force and acceleration, potential energy, kinetic energy, conservation of energy
Includes Comprehensive accessory pack to facilitate a variety of demonstrations
Weight (English) 19.8 lb.
Weight (Metric) 9kg