Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Magnetic Field Demonstrator
SCI_ED

Catalog No. S99350 Shop All Eisco Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S99350 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S99350 Supplier Eisco™ Supplier No. PH0803
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Magnetic Field Demonstrator designed to show configuration of a magnetic field in one or two dimensions.

  • Consists of a set of two transparent plastic plates, 6.0 x 3 x 0.24 in. (153 x 76 x 6 mm)
  • Each plate has a matrix of 98 small circular chambers, 0.5 x 0.3 in. (14 x 7 mm)
  • The transparent nature of the plates makes them suitable for use on an overhead projector

Specifications

Description Mounted on acrylic sheet with socket connections.
Shape Rectangular
For Use With (Application) Demonstrate configuration of a magnetic field in one or two dimensions
Grade 9 to 12
Color Clear
Product Type Magnetic Field Demonstrator
Includes Instructions, two transparent Plastic plates
Quantity 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.