Learn More
Eisco™ Motor Generator
SCI_ED
Catalog No. S96319 Shop All Eisco Products
Change view
Click to view available options
Voltage:
6 VDC
Catalog No. Voltage
S96319 6 VDC
1 options
Catalog No. S96319 Supplier Eisco™ Supplier No. PH1245N8/DEL
Includes
2-Pole armature, field coil pair, permanent magnet pair, four brushes, commutator and slip rings and instructions.
For basic training in electricity.
For experiments with AC and DC operation.
  • Dissectible device shows crucial similarities
  • Field coil and permanent magnets are on interchangeable brackets
  • Requires 6 V DC power source
Specifications
For Use With (Application) Basic training in electricity, experiments with AC and DC operation
Voltage 6 VDC
For Use With (Equipment) For experiments with AC and DC operation
Includes 2-pole armature, field coil pair, permanent magnet pair, four brushes, commutator and slip rings and instruction
Product Type Motors and Generators