Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Physics Kit: Electricity System 1
SCI_ED

Catalog No. S13471 Shop All Eisco Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13471 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13471 Supplier Eisco™ Supplier No. EIRQ08
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Students can explore common experiments such as Ohm's Law, Series and Parallel Resistance Circuits, Kirchhoff's Laws, and RC Circuits with the Electricity System 1 Physics Kit.

  • Over 50 components for building circuits
  • Use to teach 16 activities
  • Includes Activity Guides with Student worksheets

Experiments Covered

  • Simple Electric Circuit with Switches
  • Voltmeter Use
  • The Voltage Divider
  • Ammeter Use
  • The Potentiometer as a Variable Resistor
  • Ohm's Law and the Voltametric Method
  • Series Resistance Circuits
  • Parallel Resistance Circuits
  • Kirchhoff's First Law
  • Kirchhoff's Second Law
  • Series Capacitance Circuits
  • Parallel Capacitance Circuits
  • RC Circuits: Charging
  • RC Circuits: Discharging
  • The Magnetic Field of a Line of Wire
  • The Magnetic Field of a Coil

Includes: Board for Electricity and Electronics (1), Analog Voltmeter, 0 to 15 V, 0 to 1.5 V (1), Analog Ammeter, 0 to 500 mA, 0 to 50 mA (1), 100 units resistor (4), 220 Units resistor (2), 470 Units resistor (2), 1 k Units resistor (2), 4.7 k Units resistor (2), 10 k Units resistor (2), 100 Units potentiometer (1), 1000ìF capacitor (2), Lamp holder for E10 bulbs (4), Bridging Plug (4), Two cell holder (1), Single cell holder (1), 10 mH inductor (coil) (1), Compass (1), Toggle switch (1), Push button switch (1), Lamp bulb E10, 3.5 V (4), Lamp bulb E10, 6 V (4), Lamp bulb E10, 12 V (4), Cylindrical magnet (1), Ferromagnetic core (hexagonal steel screw and nut) (1), Banana plug cord, 25 cm, red (2), Banana plug cord, 25 cm, black (2), Banana plug cord, 50 cm, red (1), Banana plug cord, 50 cm, black (1)

Specifications

Grade 9 to Undergraduate
Includes Bread Board, Voltmeter, Ammeter, 6 Resistors, 2 Capacitors, Potentiometer, Lamp Holder, Bridge Plug, 2 Cell Holders, Inductor, Compass, 2 Switches, 3 Lamp Boards, Magnet, Ferromagnetic Core, Banana Plug Cord
Class Size 1 Lab Group
Product Type Electricity System 1 Physics Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.