Learn More
Eisco™ Plastic Dynamic Cart Set
SCI_ED
Catalog No. s05833 Shop All Eisco Products
Change view
Click to view available options
Pair of ABS Plastic Dynamic Carts
Catalog No.
S05833 Pair of ABS Plastic Dynamic Carts
1 options
Catalog No. S05833 Supplier Eisco™ Supplier No. PH0351PLN8
Only null left
Add to Cart
Add to Cart
Excellent upgrade from the traditional wooden or metallic carts.
Pair of single-piece, virtually unbreakable ABS carts can be used for a variety of precision experiments.
  • Body mounted on set of three low-friction nylon wheels to produce straight-line travel over long distances
  • Lockable spring-load punch has two levels of compression, activated by trigger knob on cart top
  • Cushioned cart surface supports addition of more masses to increase effective weight of cart
  • Carts are stackable to create single cart of two or three times the basic cart mass
  • Velcro™ rear pads enable study of inelastic collisions
Specifications
Product Type Dynamic Cart Set
Material Plastic
For Use With (Application) Used for a variety of precision experiments
Includes Rear pads