Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Six Hole Wood Test tube Rack
SCI_ED

Catalog No. S07854 Shop All Eisco Products
Change view
Click to view available options
Positions:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
S07854 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07854 Supplier Eisco™ Supplier No. CH0002C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Six Hole Wood Test tube Rack with coated finish to protect from chemicals.

  • Made of wood.
  • Holds six tubes with 20 mm holes.

Specifications

Autoclavable Autoclavable
Color Brown
Holds Tubes (24 mm)
Material Wood
Positions 6
Shape Vertical
Stackable Yes, When Empty
Sterility Non-sterile
Diameter (Metric) Opening 24 mm
Product Type Tube Rack
Shelves 2 Tiers
Array 6 holes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.