Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Student Cell and Porous Cup Set
SCI_ED

Catalog No. S12986 Shop All Eisco Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S12986 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12986 Supplier Eisco™ Supplier No. PH0927N8
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

One plastic student cell complete with a screw-on plastic ring, two adjustable electrode holders, porous ceramic cup and ten electrodes, one each of aluminum, nickel, tin, graphite, iron and copper and two each of lead and zinc and instructions.

The Student Cell and Porous Cup Set Student Cell and Porous Cup Set is ideal for demonstrating the characteristics of primary cells.

  • Basic voltaic student cell set is ideal for demonstrating the characteristics of primary cells
  • Set includes one plastic student cell complete with a screw-on plastic ring, two adjustable electrode holders, porous ceramic cup and ten electrodes (one each of aluminum, nickel, tin, graphite, iron and copper and two each of lead and zinc)
  • Complete instructions are included

Specifications

Grade 7 to 12
Includes One plastic student cell complete with screw-on plastic ring, two adjustable electrode holders, one porous ceramic cup and ten electrodes
Weight (English) 0.8 lb.
Weight (Metric) 0.3 kg
Class Size 1 Lab Group
Product Type Student Cell and Porous Cup Set
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.