Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EKLF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586441 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586441 Supplier Invitrogen™ Supplier No. PA586441
  • $633.00
    $492.65 / Each of 1

    Save $140.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

KLF1 is a hematopoietic-specific transcription factor that induces high-level expression of adult beta-globin and other erythroid genes. The zinc-finger protein binds to the DNA sequence CCACACCCT found in the beta hemoglobin promoter. Heterozygous loss-of-function mutations in this gene result in the dominant Inblood phenotype.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EKLF
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Klf1
Gene Accession No. P46099, Q13351
Gene Alias CDAN4; Eklf; Erythroid krueppel-like transcription factor; erythroid Kruppel-like factor; erythroid transcription factor; erythroid-specific transcription factor EKLF; HBFQTL6; INLU; KLF1; Krueppel-like factor 1; Kruppel like factor 1; Kruppel-like factor 1 (erythroid); monoclonal antibody A3D8; Nan; neonatal anemia; truncated EKLF; truncated erythroid Kruppel-like factor 1
Gene Symbols Klf1
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 288-335 of Human EKLF.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10661, 16596, 304666
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.