Learn More
Thermco Products EL-USB-1-PRO Industrial Temperature Logger
Catalog No. 04500346
Change view
Click to view available options
Software Type:
Web, Android, Apple IOS
Catalog No. Software Type
04-500-346 Web, Android, Apple IOS
1 options
Catalog No. 04-500-346 Supplier Thermco Products Supplier No. ELUSB1PRO
Add to Cart
Add to Cart
EL-USB-1-PRO Industrial Temperature Logger records data from -40°C to +125°C (-40°F to +257°F).
  • Rugged stainless steel build resists corrosion, impact, and water ingress, meeting IP67 and NEMA 4X standards.
  • Easy setup with a user-friendly, plug-and-play configuration using the included EasyLog software.
  • Extensive data analysis capabilities to visualize, print, and export data for detailed analysis.
  • High-temperature battery supplied with a specialized 2⁄3 AA battery for extended usage.
Specifications
Type Datalogger
Display Type No Display
Alarm Types Audible
Software Type Web, Android, Apple IOS