Learn More
Thermco Products EL-WiFi-VACX High-Accuracy WiFi Vaccine Monitoring Kit with Alarm Warning Light and Sounder
Catalog No. 04500331
Change view
Click to view available options
Display Type:
Status Indicator Lights
Catalog No. Display Type
04-500-331 Status Indicator Lights
1 options
Catalog No. 04-500-331 Supplier Thermco Products Supplier No. ELWIFIVACX
Add to Cart
Add to Cart
EL-WiFi-VACX High-Accuracy WiFi Vaccine Monitoring Kit has a temperature range of -40 to +60°C (-40 to 140°F).
  • The EL-WiFi-VACX features built-in sounder and LED alerts for remote vaccine monitoring and WiFi-based data logging.
  • Access data from any internet-enabled device, including smartphones, tablets, or PCs, via the EasyLog Cloud.
  • High and low alarms can be configured to send instant email notifications if fridge or freezer temperatures deviate from set limits.
  • Includes a glycol-filled probe that mimics vaccine thermal response for precise and representative temperature readings.
  • The probe is replaceable and can be calibrated independently.
  • If WiFi connectivity is lost, data is stored internally and automatically uploaded once the connection is restored.
  • EasyLog Cloud apps for Android and iOS enable simple setup and anytime, anywhere data access.
  • Exceeds CDC vaccine monitoring standards.
  • Supplied with a high-accuracy glycol probe, traceable calibration certificate, and wall bracket.
  • Complies with BS EN 12830:2018 requirements for storage and transport of temperature-sensitive goods.
Specifications
Type Temperature Datalogger
Display Type Status Indicator Lights
Alarm Types LED Indicator, E-mail