Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alkaline Phosphatase (ALP) Activity Kit

Catalog No. EEA002
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA002 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA002 Supplier Invitrogen™ Supplier No. EEA002
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Alkaline Phosphatase (ALP) Activity Kit is a highly reliable and sensitive assay designed for the quantitative measurement of ALP activity in various sample types. ALP is a group of cytomembrane-related enzymes with hydrolysis and transfer activity, acting on a variety of phosphate substrates. ALP is a homologous dimerase where each catalytic site contains three metal ions.

The assay is based on the ability of alkaline phosphatase to decompose benzene disodium phosphate to produce free phenol and phosphoric acid. Phenol reacts with 4-aminopyrine in alkaline solution and oxidizes with potassium ferricyanide to form a red quinone derivative. The enzyme activity can be calculated indirectly by measuring the OD value.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.13–50 King Units/100 mL
Assay Sensitivity 0.13 King Units/100 mL
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Plasma, Serum, Tissue Homogenates, Whole Cells
For Use With (Equipment) Microplate Reader
Incubation Time 15 min.
Interassay CV 8.5
Intraassay CV 5.1
Quantity 96 tests
Sample Volume 5 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Alkaline Phosphatase (ALP)
Total Assay Time 30 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.