Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cell Copper (Cu) Colorimetric Assay Kit

Catalog No. EEA007
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA007 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA007 Supplier Invitrogen™ Supplier No. EEA007
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Cell Copper Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of copper ion content in cells.

In this assay, under acidic conditions, copper ions in the sample react with a complexing agent to form a purple complex that has a maximum absorption peak at 580 nm. The copper ion content can be calculated indirectly by measuring the OD value at 580 nm.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.18–5 μmol/L
Assay Sensitivity 0.18 μmol/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Whole Cells
For Use With (Equipment) Microplate Reader
Incubation Time 5 min.
Interassay CV 3.1
Intraassay CV 3
Quantity 96 tests
Sample Volume 100 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Cell Copper (Cu)
Total Assay Time 10 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.