Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ γ-Glutamyl Transferase (GGT/γ-GT) Colorimetric Activity Kit

Catalog No. EEA029
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA029 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA029 Supplier Invitrogen™ Supplier No. EEA029
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The γ-Glutamyl Transferase (GGT/γ-GT) Colorimetric Activity Kit is a highly reliable and sensitive assay designed for the quantitative measurement of GGT in serum, plasma, and animal tissue samples. GGT is widely present in various organs of the human body, including the liver, kidney, and pancreas. GGT catalyzes the degradation of glutathione (GSH) and is involved in the regulation of glutathione levels in tissues, as well as the absorption and excretion of amino acids. Changes in GGT activity have been associated with various diseases, particularly those affecting the hepatobiliary system.

The assay utilizes the catalytic property of GGT to transfer a γ-glutamyl group from glutamyl p-nitroaniline to N-glycyl glycine, resulting in the production of p-nitroaniline. The p-nitroaniline has a characteristic absorption peak at 405 nm, allowing for the measurement of GGT activity.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.88–399.4 U/L
Assay Sensitivity 0.88 U/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Microplate Reader
Incubation Time 6 min.
Interassay CV 6.2
Intraassay CV 4.2
Quantity 96 tests
Sample Volume 25 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named γ-Glutamyl Transferase (GGT/γ-GT)
Total Assay Time 20 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.