Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lactate Dehydrogenase (LDH) Colorimetric Activity Kit

Catalog No. EEA013
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA013 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA013 Supplier Invitrogen™ Supplier No. EEA013
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Lactate Dehydrogenase (LDH) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of LDH in serum, plasma, tissue, and cell culture samples. LDH is an enzyme that catalyzes the conversion of lactate to pyruvate, an essential step in cellular energy metabolism. Elevated LDH levels are associated with tissue damage, inflammation, and certain diseases, making it a useful biomarker in clinical and research settings.

In this assay, LDH catalyzes lactic acid to produce pyruvate. Pyruvate reacts with 2, 4-dinitrophenylhydrazine to form pyruvate dinitrophenylhydrazone, which is red-brown in alkaline solution. The color intensity is proportional to pyruvate concentration, where the activity of LDH can be calculated by measuring OD value.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 6–1000 U/L
Assay Sensitivity 6 U/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Hydrothorax, Plasma, Serum, Tissue Homogenates, Whole Cells
For Use With (Equipment) Microplate Reader
Incubation Time 35 min.
Interassay CV 2.4
Intraassay CV 1.8
Quantity 96 tests
Sample Volume 40 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Lactate Dehydrogenase (LDH)
Total Assay Time 45 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.