Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Malondialdehyde (MDA) Colorimetric Assay Kit

Catalog No. EEA015
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA015 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA015 Supplier Invitrogen™ Supplier No. EEA015
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Malondialdehyde (MDA) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of MDA in serum, plasma, tissue, and other samples. MDA is a biomarker of oxidative stress and lipid peroxidation, and detection of MDA in samples can indirectly reflect the level of cellular damage from oxygen free radicals.

In this assay MDA, as the catabolite of lipid peroxide, reacts with thiobarbituric acid (TBA) to produce a red compound. This compound has a maximum absorption peak at 532 nm, which can be used to measure the amount of malondialdehyde present.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 2.92–40 μmol/L
Assay Sensitivity 1.13 μmol/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Microplate Reader
Incubation Time 40 min.
Interassay CV 7.2
Intraassay CV 4.1
Quantity 96 tests
Sample Volume 40 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Malondialdehyde (MDA)
Total Assay Time 65 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.