Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit (FRAP method)

Catalog No. EEA024
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA024 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA024 Supplier Invitrogen™ Supplier No. EEA024
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit (FRAP method) is a highly reliable and sensitive assay designed for the quantitative measurement of T-AOC in serum, plasma, tissues, cells, cell culture supernatant, saliva, and urine sample types. T-AOC refers to the overall ability of a biological sample to counteract oxidative stress by neutralizing reactive oxygen species (ROS) and other harmful free radicals. It represents the cumulative effect of both enzymatic and non-enzymatic antioxidants present in the sample.

The assay quantifies the ability of the sample to reduce ferric ions (Fe3+) to ferrous ions (Fe2+) using the ferric reducing antioxidant power (FRAP) method under acidic conditions. T-AOC can be measured using the absorbance value of the resulting blue solution at 593 nm.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.23–2 mmol Trolox Equiv./L
Assay Sensitivity 0.049 mmol/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Cell Culture Supernatant, Plasma, Saliva, Serum, Tissue Homogenates, Urine, Whole Cells
For Use With (Equipment) Microplate Reader
Incubation Time 3-5 min.
Interassay CV 8.1
Intraassay CV 3.9
Quantity 96 tests
Sample Volume 5 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Total Antioxidant Capacity (T-AOC)
Total Assay Time 20 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.