Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Total Cholesterol (TC) Colorimetric Assay Kit (single reagent, COD-PAP method)

Catalog No. EEA026
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEA026 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEA026 Supplier Invitrogen™ Supplier No. EEA026
Add to Cart
Add to Cart

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Total Cholesterol (TC) Colorimetric Assay Kit (single reagent, COD-PAP method) is a highly reliable and sensitive assay designed for the quantitative measurement of total cholesterol in serum, plasma, and animal tissue. Cholesterol is a type of sterol and lipid found in cell membranes. It is an essential component of cell structure and function, but high levels of TC in the blood have been linked to an increased risk of cardiovascular disease. The measurement of TC levels (which includes free cholesterol and cholesterol esters) is an important diagnostic tool in clinical and research settings.

The assay utilizes the COD-PAP method, which involves the hydrolysis of cholesterol esters in the sample to produce free cholesterol and free fatty acids. Free cholesterol is then oxidized by cholesterol oxidase to produce 4-cholestenone and hydrogen peroxide. In the presence of 4-aminoamylpyridine and phenol, hydrogen peroxide catalyzes peroxidase to form red quinone compounds. The color depth of the generated quinone is directly proportional to the cholesterol content, which can be measured spectrophotometrically at 510 nm.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.29 to 25.85 mmol/L
Assay Sensitivity 0.29 mmol/L
Content And Storage Store at 2°C to 8°C.
Detection Method Colorimetric
Sample Type Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Microplate Reader
Incubation Time 10 min.
Interassay CV 8.3
Intraassay CV 3.1
Quantity 96 tests
Sample Volume 2.5 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Total Cholesterol (TC)
Total Assay Time 20 min.
Show More Show Less
Can I use the Total Cholesterol (TC) Colorimetric Assay Kit (single reagent, COD-PAP method) (Cat. No. EEA026) with cell samples?

We do not recommend using the Total Cholesterol (TC) Colorimetric Assay Kit with cell samples because most untreated cell samples have a low TC that does not meet the sensitivity of the kit.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.