Learn More
Selleck Chemical LLC Elafibranor S3720-5mg
SDP
Supplier:  Selleck Chemical LLC S37205MG
Encompass_Preferred
Elafibranor (GFT505) is an agonist of the peroxisome proliferator-activated receptor- (PPAR-alpha) and peroxisome proliferator-activated receptor- (PPAR- ) It improves insulin sensitivity glucose homeostasis and lipid metabolism and reduces inflammation
Catalog No. 50-194-8234
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.