Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Elastase, Human Neutrophil, Protein

Catalog No. 32468150UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-468-150UG 50 μg
32-468-1100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-468-150UG Supplier MilliporeSigma™ Supplier No. 32468150UG
Add to Cart
Add to Cart
Native elastase from human neutrophil. A serine protease that degrades elastin, collagen, and proteoglycans. Its overexpression has been implicated in the development of pulmonary emphysema, rheumatoid arthritis, and cystic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Salt-free.
Preparation Method Reconstitute in 50mM sodium acetate, pH 5.5, with 200mM NaCl or in other aqueous buffers. Material is most active near a neutral pH, but to maintain stability upon reconstitution, use in a slightly acidic buffer (pH 5.5) to avoid autolysis.
Purification Method Major band by SDS-PAGE
Quantity 50 μg
Immunogen Elastase
Storage Requirements Following reconstitution, aliquot and freeze (−20°C) for long term storage or refrigerate (4°C) for short-term storage. Acidic stock solutions are stable for up to 1 month at 4°C or for up to 1 year at −20°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.