Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Elastase, porcine pancreas

Catalog No. AAJ6187403
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6187403 1 g
AAJ61874MB 25 mg
AAJ61874MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6187403 Supplier Thermo Scientific Chemicals Supplier No. J6187403
Add to Cart

Elastase, porcine pancreas, CAS # 39445-21-1, is a single polypeptide chain of 240 amino acid residues isolated from the porcine pancreas. It is a serine protease that degrades elastin, a matrix component of tissues. | CAS: 39445-21-125 kd g/mol

This enzyme can be used alone or in combination with other proteases for protein analysis by mass spectrometry and other applications. Elastase, is used in tissue dissociation because elastin is found in highest concentrations in the elastic fibers of connective tissues, elastase is frequently used to dissociate tissues that contain extensive intercellular fiber networks. For this purpose, it is usually used with other enzymes such as collagenase, trypsin, and chymotrypsin, membrane protein solubilization, protein sequence studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Elastase is a serine protease that can hydrolyze elastin, a specific protein of elastic fibers

Application

• Elastase, porcine pancreas has been used in cell culture techniques, ELISA assays, and in experiments to measure the inhibitory activity of elastase

• Since elastin is found in the fibers of connective tissues, elastase is commonly used to dissociate tissues that contain extensive intercellular fiber networks

• It is typically used with other enzymes, including collagenase, trypsin, and chymotrypsin

• Elastase enzyme is isolated from porcine pancreas and can be used alone or combined with other proteases for protein analysis by mass spectrometry and other applications, including membrane protein solubilization and protein sequence studies

• This compound also hydrolyzes amides and esters such as N-Benzoyl-L-alanine methyl ester

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39445-21-1
Synonym E.C. 3.4.21.36; Pancreatopeptidase E

Specifications

For Use With (Application) A serine protease that acts on elastin
Molecular Weight (g/mol) 25 kd
Content And Storage 2°C to 8°C
Form Lyophilized powder,MW 25000
Quantity 1 g
Solubility Soluble in water or Tris buffer up to 2.5mg/ml; Avoid high salt concentrations (ie >50mM) if salt is necessary in the buffer; Unstable in solution at pH <3.5
Product Type Elastase
EINECSNumber 254-453-6
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.