Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Elastase, Porcine Pancreas, High Purity, Crystallized, Protein

Catalog No. 324682250U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1000 U
250 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-468-2250U 250 U
32-468-21000U 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-468-2250U Supplier MilliporeSigma™ Supplier No. 324682250U
Add to Cart
Add to Cart
Native elastase from porcine pancreas. A serine protase that catalyzes the hydrolysis of proteins and peptides (especially at bonds adjacent to neutral amino acid residues), including albumin, casein, denatured collagen, elastin, fibrin, and hemoglobin and of a number of synthetic substrates containing aspartic acid, phenylalanine, or tyrosine. Inhibited by DFP, elastinal, and α2-macroglobulin.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 50mM Trehalose, 1mM Acetic Acid.
Preparation Method dH2O (10mg/mL)
Quantity 250 U
Immunogen Elastase
Storage Requirements Following reconstitution, elastase may be stored at 4°C at pH 6.0 for long term use. If incubated at room temperature at or near its optimum pH, elastase rapidly autolyzes to a mixture of peptides. Stock solutions are stable for up to 2 months at 4°C, pH 6.0.
Regulatory Status RUO
Species Porcine
Recombinant Native
Form Lyophilized
Purity or Quality Grade High Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.