Description
- Get the complete safety package – unlike signs and tapes this cover provides a sign and an unavoidable boundary in one easy-to-use product
- Elastic covers are ideal for quick protection needs and provide ventilation for areas that require it
- Make it last – unlike wasteful warning tape, these covers are designed to be reused for multiple projects
Specifications
Specifications
| Product Type | Confined Space Cover |
| Color | Red |
| Material | Polyester |
| Diameter (English) | 36 in. |
| Displayed Text | Permit required. No unauthorized entry. |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.