Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Elastin Monoclonal Antibody (BA-4)
GREENER_CHOICE

Catalog No. MA127129 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA127129 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA127129 Supplier Invitrogen™ Supplier No. MA127129
  • $667.50
    $506.65 / Each of 1

    Save $160.85

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Proteolytic pretreatment is required for immunostaining of paraffin embeded tissue. The positive control of Heart tissue is suggested.

This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Elastin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone BA-4
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.09% sodium azide
Gene ELN
Gene Accession No. P04985, P15502
Gene Alias 5830419M17Rik; AI385707; AI480567; AW229038; BB105748; E030024M20Rik; elastin; elastin (supravalvular aortic stenosis, Williams-Beuren syndrome); elastin a precursor; elastin b precursor; elastin c precursor; elastin microfibril interface located protein 1; elastin microfibril interface-located protein 1; Elastin microfibril interfacer 1; Eln; EMI; EMILIN; EMILIN1; EMILIN-1; FLJ38671; FLJ43523; gp115; RATTREL11; SVAS; TREL11; Trela; Trela26; Tropoelastin; WBS; WS
Gene Symbols ELN
Host Species Mouse
Immunogen Bovine alpha-Elastin.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2006, 280781
Target Species Bovine, Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.