Learn More
Sartorius Elbow Pad
Catalog No. 14559559 Shop All Sartorius Products
Change view
Click to view available options
Visco elastic elbow pad
Catalog No.
14-559-559 Visco elastic elbow pad
1 options
Catalog No. 14-559-559 Supplier Sartorius Supplier No. 723103
Relieves contact stress, pain and discomfort underneath elbow while pipetting
  • Improves pipetting ergonomics
  • Forms according to any elbow size or shape
  • Coating is pleasant to the skin
  • Compact size: requires little desk space
  • Durable
  • Easy to clean: Can be cleaned using washing up liquid, ethanol (70%) or Biocontrol decontamination solution
  • Non-autoclavable
  • Can be used for long pipetting series, work requiring high concentration, e.g. micro plate work, any work where a cushion underneath elbow or wrist is needed
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
For Use With Pipettes
Material Visco Elastic
For Use With (Equipment) Microplate work or micro volume pipetteting
WARNING: Cancer - www.P65Warnings.ca.gov