Learn More
Grainger Elct Flr Htr Thrmostat 4-1/2 H 2 Pole
Supplier:  Grainger 14C467
Encompass_Preferred
The ThermoSoft Electric Floor Heater Thermostat is a programmable thermostat designed for use with electric floor heating systems. It features a large LCD display with backlighting and intuitive programming for setting customized heating schedules. The thermostat provides floor temperature sensing and control, with a temperature range of 41°F to 104°F. It is compatible with 120V and 240V heaters up to 15A and offers adjustable differential/cycle rates. The unit can be surface mounted on a wall and includes a floor sensor probe with 25 ft cable.
Catalog No. 50-275-2322
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.