Learn More
Grainger Electric Drill 3/8 In 0 to 2500 rpm 4.9A
Supplier:  Grainger 14G994
Encompass_Preferred
The Grainger Approved 3/8 in. Variable Speed Reversible Electric Drill features a powerful 7 amp motor that delivers 0-2,500 RPM. It has an all-metal keyless chuck with 24-position clutch to prevent overtightening. The drill offers 2-speed gearing for optimal torque and control. It has a soft grip handle for comfort and control. The electric drill comes with a cord wrap for organization. This is a versatile, heavy-duty electric drill suitable for drilling, driving and light-duty hammer applications.
Catalog No. 50-275-1003
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.