Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Electric Pipette Controller, 0.1 to 200 mL
SCI_ED

Catalog No. p-7225992 Shop All Walter Products Products
Change view
Click to view available options
Color:
Dark Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S139025 Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139025 Supplier Walter Products™ Supplier No. TEL01
Only null left
Add to Cart
Add to Cart

These motorized pipette fillers are easy-to-use and attach to both glass and plastic pipettes ranging from 0.1 to 200 mL.

  • Single-handed operation for both right and left handed operators
  • Fits both glass and plastic pipettes from 0.1 to 200 mL
  • Safety valve and hydrophobic membrane filter provide double protection against fluid penetration
  • At maximum motor speed, a 50 mL pipette is filled in less than 10 s
  • Approximately 12 hours of continuous pipetting with a 10 mL pipette without recharging

Specifications

Color Dark Blue
Volume (Metric) 200 mL
For Use With (Application) Pipette Filling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.