Learn More
Med Vet International Replacement Defibrillator Pads Cartridge Philips HeartStart Adult, M5071A, Each
Supplier:  Med Vet International 918919EA
Encompass_Preferred
HeartStart Adult SMART Pads are appropriate for cardiac arrest victims weighing 55lbs or more Pads should be replaced after each use, or once every (2) years Cartridge can be easily snapped into the OnSite AED, making replacing your electrodes easier than ever Convenient "Pull" handle makes access easy and time efficient Holds one set of adult electrode pads
Catalog No. 50-304-4106
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.