Learn More
Cole-Parmer Cole-Parmer General Purpose pH Electrode, 100 Ohms Pt RTD, 10-ft Cable
Supplier: Cole-Parmer 0599426
General-purpose single-junction electrodes are economical yet accurate. Can be used for measurements up to pH 13. Maximum pressure is 150 psi at 25°>C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.