Learn More
BINDER™ Electromechanical Door Lock with Pin
SDP
Catalog No. 502875739 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB 65, KB 130, KB 260, KB 470
KB 720, KB 1060
KB PRO 130/260/470
KB PRO 260, KBF PRO 130/260/470
KB PRO 720/1060/1600
KBF 130, KBF 260, KBF 470
KBF 720, KBF 1060, KBF 1600
KBF PRO 720/1060/1600
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5739 KB 65, KB 130, KB 260, KB 470
50-287-5698 KB PRO 130/260/470
50-287-5760 KBF 130, KBF 260, KBF 470
50-287-5592 KB PRO 260, KBF PRO 130/260/470
50-287-5743 KB 720, KB 1060
50-287-5626 KB PRO 720/1060/1600
50-287-5578 KBF 720, KBF 1060, KBF 1600
50-287-5627 KBF PRO 720/1060/1600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 50-287-5739 Supplier BINDER™ Supplier No. 80122531
Add to Cart
Add to Cart
Lockable door handle that can be opened with PIN code via controller
Door is locked with a PIN code via a controller to ensure secure access to samples.
Specifications
For Use With (Equipment) KB 65, KB 130, KB 260, KB 470
Material Steel
Type Locking Latch