Learn More
Thermo Scientific™ Electronic Hand-Held Crimper/Decrimper
Catalog No. 03254050 Shop All Thermo Scientific Products
Change view
Click to view available options
Size (Metric):
20 mm
Catalog No. Size (Metric)
03-254-050 20 mm
1 options
Catalog No. 03-254-050 Supplier Thermo Scientific™ Supplier No. 60180ECR20FO
Add to Cart
Add to Cart
Use Thermo Scientific™ Electronic Hand-Held Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively. For Level 3 High Performance Applications.
Use Thermo Scientific™ Electronic Hand-Held Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively. These hand-held, high-power crimpers and decrimpers allow you to seal or remove crimp caps with a simple, one-handed push of a button. Use for crimping of vials with aluminum, pressure-release, and bi-metal caps.
Features:
  • Hand-Held Crimper and Decrimper
  • The battery charge icon on the color screen changes to red when battery is low
  • Includes a fully rechargeable lithium ion battery and a universal power supply/recharger with international plug adapters
  • Adjustable settings for best crimp results and a motor with extended lifetime and no particle emission.
  • The message window conveniently provides all-important information at one glance.
  • Performance Level 3, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.
The Message Window Displays:
  • The actual crimp power
  • An error message after an incorrect crimping process
  • Statistics and log of recent crimping results
  • In the language of your choice (English, Chinese, Japanese, German, French, Spanish, Italian, Korean)
TRUSTED_SUSTAINABILITY
Specifications
Type Electronic Crimper
Size (Metric) 20 mm
For Use With (Equipment) Crimp Cap