Learn More
Chemglass Life Sciences Electrophoresis System, Complete
SDP
Supplier:  Chemglass Life Sciences CLS190701
Encompass
Electrophoresis system equipped with blue light LEDs to allow for immediate visualization of DNA/RNA bands while running agarose gels. The unit comes complete with three gel trays, combs, and gel casting dams. The power supply is built in. This safe system is safe, unique, and powerful. Gel box can be easily removed from base illuminator. The box can only be placed one way onto the base to avoid running the power incorrectly.
Catalog No. 50-153-2250
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.