Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Electroscopes Kit
SDPSCI_ED

Catalog No. S139485 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139485 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139485 Supplier United Scientific™ Supplier No. ELKIT1
Add to Cart
Add to Cart

Demonstrate key electrostatics concepts with this kit.

Demonstrate key electrostatics concepts with this kit. Contains everything necessary to construct two flask form electroscopes. Detailed manual included experiments using two electroscopes for positive and negative charge studies, and a Faraday's Pail experiment.

Contents:

  • 2 glass flasks
  • 2 aluminum disc terminals
  • 2 sets of 4 foil leaves
  • 2 electrode assemblies
  • 1 aluminum can (Faraday pail)
  • 1 hard rubber rod
  • 1 silk cloth
  • 1 acrylic rod
  • 1 wool cloth

Specifications

Grade 5 to 12
Format Kit
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.