Learn More
Description
Includes
Two electroscopes with flasks; two ball and disc terminals; Faraday cage; six friction rods, labeled; electrophorus with charge plate and handle; neon lamp; ice pail; acetate and polyethylene cloth; charge transfer ball; conductive ball; pith balls; mounted point; instructions
- Innovative, economical kit
- Teach the basics of circuits and logic board
- Contains everything needed to create series and parallel circuits, switches, fuses, light bulbs and more
- All the components attach easily to the injection-molded base
- With illustrated, step-by-step instructions
Specifications
Specifications
| For Use With (Application) | Teach the basics of circuits and logic board |
| Product Type | Electrostatic Demo Kit |
| Includes | 2 Electroscopes with flasks, 2 ball and disc terminals, Faraday cage, 6 friction rods, electrophorus with charge plate and handle, Neon lamp, ice pail, acetate and Polyethylene cloth, charge transfer ball, conductive ball, pith balls, mounted point instructions |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

