Learn More
Vernier Electrostatics Kit
SDPSCI_ED
Catalog No. S40821 Shop All Vernier Software Products
Change view
Click to view available options
Grade:
College and High School
Catalog No. Grade
S40821 College and High School
1 options
Catalog No. S40821 Supplier Vernier Supplier No. ESKCRG
Add to Cart
Add to Cart
The Electrostatics Kit allows students to perform a range of experiments in electrostatics with the Vernier Charge Sensor.
The Electrostatics Kit is an accessory for the Vernier Charge Sensor. This kit allows students to perform a range of experiments in electrostatics including the use of Faraday’s pail, quantitative and qualitative measurement of charge, charging by friction, charging by contact and charging by induction.
Specifications
Includes Faraday pail and cageGrounding planeGrounding wires and wrist strapCharge producers and proof planeWool, vinyl, nylon rod, PVC rodCotton cloth
Grade College and High School
WARNING: Cancer - www.P65Warnings.ca.gov