Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Electrostatics Kit
SDPSCI_ED

Catalog No. S27799 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S27799 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27799 Supplier United Scientific Supplies Supplier No. ELSKIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kit is a collection of materials designed for investigation of creation of charges through various rods and fabrics

Specifications

Grade 4 to 12
Age 10 to 18
Includes Hollow Glass Friction Rod (1), Glass Friction Rod, Half Ground (1), Ebonite Rod (1), String (10 ft.), Balloons (3), Silk Pad (6 in.), Faux Fur Pad (6 in.), and Flannel Pad (6 in.)
Class Size 1 Lab Group
Product Type Electrostatics Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.