Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Elementary Simple Machines Kit
SDPSCI_ED

Catalog No. S96414 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
3 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S96414 3 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96414 Supplier United Scientific Supplies Supplier No. FSMKIT02E
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Set contains: lever with centimeter scale, two pulleys, spring scale, two weight hangers, slotted weight set, base and support rod, support rod with clamp, two hook collars, pulley cord, and manual.

Ideal for introducing students to the concepts of changing force by levers and pulleys, and the notion of mechanical advantage.

  • Manual describes experiments that can be performed with the three types of levers and single and double pulley systems
  • Also shows the characteristics of each machine

Specifications

Product Type Kit, Elementary Simple Machines
Grade 3 to 8
For Use With (Application) Introducing students to the concepts of changing force by levers and pulleys and the notion of mechanical advantage
Includes Set contains: lever with centimeter scale, two pulleys, spring scale, two weight hangers, slotted weight set, base and support rod, support rod with clamp, two hook collars, pulley cord and manual
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.