Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Elements™ EM3 Polyurethane-Coated Cut-Resistant Gloves

Catalog No. 17200419 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-419 Large
17-200-417 Small
17-200-420 X-Large
17-200-421 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-200-419 Supplier Wells Lamont™ Supplier No. EM3L
Add to Cart
Add to Cart

Elements™ EM3 Polyurethane-Coated Cut-Resistant knit gloves feature a waterproof membrane and polyurethane coating on the fingertips for easy use on tablets or smartphones.

  • Combines cut resistance with a waterproof membrane and nylon liner for comfort and protection
  • Membrane is constructed of a blend of nylon, metal wire and HPPE
  • Polyurethane coating on the fingertips for easy use on tablets and smartphones
  • Gloves provide ANSI Abrasion Level 2, and Cut Level A3 protection
  • Reduces the need for multiple disposable products

Specifications

ANSI Abrasion Rating ANSI 2
Construction Knit
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A3
Hand Specific
Material HPPE, Nylon, Metal
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Machining, Oily Parts Assembly, and Small Parts Handling
Glove Cuff Style Knit
Liner Material Nylon
Model EM3L
Puncture Rating ANSI/ISEA 105-16 Level 3
Series EM3
Yarn Gauge 18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.