Learn More
Mopec Elevating Pedestal Style Trimming Table
Features a down draft ventilation system, which helps ventilate during a necropsy. Mopec™ Elevating Pedestal Style Trimming Table incorporates an elevating mechanism within the pedestal, capable of lifting and lowering 8 inches.
Supplier: Mopec OC200
Description
- Stainless steel elevating pedestal necropsy table with downdraft ventilation
- Tabletop is 14 gauge stainless steel with #4 satin finish
- Four removable perforated grid plates.
- Hydro aspirator with built-in vacuum breaker
- Built-in large integral sink
- Hot and cold single lever mixing valve with swing spout faucet complete with vacuum breaker
- Deck mounted chrome plated brass sink spray with high pressure hose
Warranty and Services
1 Year Warranty
Specifications
Trimming Table | |
Trimming Tables | |
100 in. | |
40.5 in. | |
94 cm | |
82.5 cm | |
750 lb. | |
76.2 cm |
2 GFCI Receptacles positioned on each side of Table | |
37 in. | |
30 in. | |
32.5 in. | |
102.8 cm | |
254 cm | |
340 Kg |