Learn More
Med Vet International Miltex Root Tip Elevator, Size 79, German, Del79
Supplier:  Med Vet International MILDEL79
Encompass_Preferred
The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Root Tip Elevator. Elevators are instruments used in dental extractions or surgical procedures. In dental procedures they may be used to loosen teeth prior to forcep extraction, to remove roots or impacted teeth, when teeth are compromised and susceptible to fracture or when they are malpositioned and cannot be reached with forceps. In surgical procedures they can be used for scraping, elevating or dissecting bones or tissues. They are made from premium-grade German stainless steel. Specifications: Size 79, DEL79
Catalog No. 50-287-5576
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.